• USA Home
  • 95296 - Hydrogen ionophore IV

EMAIL THIS PAGE TO A FRIEND
95296 Sigma-Aldrich

Hydrogen ionophore IV

Selectophore, function tested

Synonym: ETH 1778, Octadecyl isonicotinate, Proton ionophore IV

  • CAS Number 103225-02-1

  • Empirical Formula (Hill Notation) C24H41NO2

  • Molecular Weight 375.59

  •  Beilstein Registry Number 5969390

  •  MDL number MFCD00144461

  •  PubChem Substance ID 329770437

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   VXDJNEIVBJLMMZ-UHFFFAOYSA-N

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Neutral hydrogen ion ionophore for solvent polymeric membrane electrodes, used in pH sensors e.g. for gastroenterological applications

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

95291 Hydrogen ionophore I - cocktail A, Selectophore
95292 Hydrogen ionophore I, Selectophore, function tested
95293 Hydrogen ionophore I - cocktail B, Selectophore
95295 Hydrogen ionophore II, Selectophore
95298 Hydrogen ionophore III, Selectophore, ≥99.0%
16979 Hydrogen ionophore V, Selectophore, function tested
43538 Tridodecylamine hydrochloride, Selectophore, ≥98.0% (AT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?