EMAIL THIS PAGE TO A FRIEND
95310 Sigma-Aldrich

L-(+)-Tartaric acid

ReagentPlus®, ≥99.5%

Synonym: (2R,3R)-(+)-Tartaric acid, L-Threaric acid

  • CAS Number 87-69-4

  • Linear Formula HO2CCH(OH)CH(OH)CO2H

  • Molecular Weight 150.09

  •  Beilstein Registry Number 1725147

  •  EC Number 201-766-0

  •  MDL number MFCD00064207

Purchase

Properties

Related Categories Acids, Acids & Bases, Asymmetric Synthesis, Biochemicals and Reagents, Carboxylic Acids,
grade   ReagentPlus®
vapor density   5.18 (vs air)
InChI Key   FEWJPZIEWOKRBE-JCYAYHJZSA-N
assay   ≥99.5%
optical activity   [α]20/D +13.5±0.5°, c = 10% in H2O
autoignition temp.   797 °F
ign. residue   ≤0.1% (as SO4)
loss   ≤0.2% loss on drying, 110 °C
mp   166-170 °C
  170-172 °C(lit.)
solubility   water: soluble150 g/L at 20 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WW7875000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?