• USA Home
  • 95325 - Micro particles based on melamine resin, nile blue-marked

EMAIL THIS PAGE TO A FRIEND
95325 Sigma-Aldrich

Micro particles based on melamine resin, nile blue-marked

size: 9 μm

Synonym: Melamine-formaldehyde resin

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Microparticles, Micro particles based on melamine resin, nile blue labeled, Microparticles
form   aqueous suspension
  particles
concentration   2.5% (solids)
particle size   9 μm std dev <0.15 μm, coeff var <2%
fluorescence   λex 633 nm; λem 672 nm
format   neat

Description

Analysis Note

For every lot, exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?