• USA Home
  • 95366 - (−)-Dimethyl D-tartrate

EMAIL THIS PAGE TO A FRIEND
95366 Sigma-Aldrich

(−)-Dimethyl D-tartrate

purum, ≥99.0% (sum of enantiomers, GC)

Synonym: D-(−)-Tartaric acid dimethyl ester, Dimethyl D-tartrate

  • CAS Number 13171-64-7

  • Linear Formula [-CH(OH)CO2CH3]2

  • Molecular Weight 178.14

  •  Beilstein Registry Number 1726254

  •  EC Number 236-118-6

  •  MDL number MFCD00008445

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Catalysts, Ligands, and Reagents, Epoxidation,
InChI Key   PVRATXCXJDHJJN-IMJSIDKUSA-N
grade   purum
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D −21±1°, c = 1% in H2O
bp   158 °C/0.2 mmHg(lit.)
mp   57-60 °C (dec.)(lit.)
  ~58 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

242942 (−)-Dimethyl D-tartrate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?