EMAIL THIS PAGE TO A FRIEND
95369 Sigma-Aldrich

Paraffin wax

extra-low viscosity

Purchase

Properties

Related Categories Core Bioreagents, Lipids and Related Products, Molecular Biology, Optimization Reagents, Protein Structural Analysis,
form   extra-low viscosity
refractive index   n20/D 1.45
viscosity   ~5 mPa.s, neat(20 °C)
mp   43-95 °C
density   0.82 g/mL at 20 °C

Description

General description

extra-light

Price and Availability


Laboratory Gloves

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
RV0350000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?