• USA Home
  • 95375 - Bismuth standard for AAS

EMAIL THIS PAGE TO A FRIEND
95375 Sigma-Aldrich

Bismuth standard for AAS

ready-to-use, traceable to BAM, in nitric acid

Synonym: Bismuth(III) nitrate solution

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Single Solution,
InChI Key   JCXGWMGPZLAOME-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   in nitric acid
  1.000 g/L Bi
application(s)   ICP/AAS: suitable
format   single component solution

Description

Preparation Note

prepared with Bi(NO3)3 ⋅ 5H2O and HNO3 (~0.5 M)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

76668 Bismuth Standard for AAS, TraceCERT®, 1000 mg/L Bi in nitric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?