• USA Home
  • 95386 - Bismuth(III) salicylate basic

EMAIL THIS PAGE TO A FRIEND
95386 Sigma-Aldrich

Bismuth(III) salicylate basic

purum, ≥97.0% (KT)

Purchase

Properties

Related Categories Bismuth Salts, Essential Chemicals, Materials Science, Metal and Ceramic Science, Research Essentials,
InChI Key   ZREIPSZUJIFJNP-UHFFFAOYSA-K
grade   purum
assay   ≥97.0% (KT)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

480789 Bismuth(III) subsalicylate, 99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?