• USA Home
  • 95538 - Micro particles based on melamine resin

EMAIL THIS PAGE TO A FRIEND
95538 Sigma-Aldrich

Micro particles based on melamine resin

analytical standard, size: 12 μm

Synonym: Melamine-formaldehyde resin, Size standards

Purchase

Properties

Related Categories Biochemicals and Reagents, Microparticles, Microparticles based on melamine resin More...
grade   analytical standard
form   aqueous suspension
  particles
concentration   10% (solids)
particle size   12 μm std dev <0.2 μm, coeff var <2%
format   neat

Description

Analysis Note

For every lot, exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer.
Density: 1.51 g/cm3
Heat resistance <300 °C
Chemical resistance: acids, bases, organic solvents.
Monodispers melamin resin particles are characterized by high uniformity and narrow size distribution. They carry functional methylol groups, which can be used for covalent coupling of biological active compounds.
Additional advantages of melamin resin particles are:
• hydrophilic surfaces
• high degree of cross linking and high pressure resistance
• long term stability in aqueous media (freezing and thawing does not damage them)
• lyophilization and resuspension in aqueous media does not damage them (powders are available on request)
• no swelling in organic solvents

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?