95681 Sigma-Aldrich

Activated Charcoal Norit®

Norit SA2, from peat, steam activated, powder

DOWNLOAD MSDS (PDF)

Synonym: Charcoal activated Norit®

  • CAS Number 7440-44-0

  • Empirical Formula (Hill Notation) C

  • Molecular Weight 12.01

  •  Beilstein Registry Number 4360473

  •  EC Number 231-153-3

  •  MDL number MFCD00133992

  •  PubChem Substance ID 24890186

Properties

Related Categories Activated Charcoal, Activated and Mesoporous Carbon, Adsorbents, Filter Aids and Drying Agents, Alternative Energy, Essential Chemicals,
biological source   from peat
form   powder
quality   steam activated
mfr. no.   Norit SA2

Description

Application

for decolorizing and purification of process liquids in biotechnology and food technology

Legal Information

Norit is a registered trademark of Norit N.V.

Price and Availability

Customers Also Viewed

Activated charcoal

decolorizing

Activated charcoal

DARCO®, −100 mesh particle size, powder

Activated Charcoal Norit<SUP>®</SUP>

Norit CA1, from wood, chemically activated, powder

Activated Charcoal Norit<SUP>®</SUP>

Norit SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder

Activated Charcoal Norit<SUP>®</SUP>

Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?