• USA Home
  • 96152 - Sulfur standard for ICP

EMAIL THIS PAGE TO A FRIEND
96152 Sigma-Aldrich

Sulfur standard for ICP

for ICP, ready-to-use, in H2O

Synonym: Ammonium sulfate solution

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, ICP-OES/-MS, Matrix Selection,
grade   for ICP
InChI Key   BFNBIHQBYMNNAN-UHFFFAOYSA-N
CofA   the measured values of the metal traces are reported on Certificate of Analysis
shelf life   limited shelf life, expiry date on the label
concentration   in H2O
  1.000 g/L S

Description

Other Notes

Aqueous ammonium sulfate solution equivalent to indicated ammonia levels

Preparation Note

prepared with H(NH4)2.SO4 and H2O

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?