EMAIL THIS PAGE TO A FRIEND
96330 Sigma-Aldrich

Cinnamyl alcohol

purum, ≥97.0% (GC)

Synonym: 3-Phenyl-2-propen-1-ol

  • CAS Number 104-54-1

  • Linear Formula C6H5CH=CHCH2OH

  • Molecular Weight 134.18

  •  Beilstein Registry Number 1903999

  •  EC Number 203-212-3

  •  MDL number MFCD00002921

  •  PubChem Substance ID 329770496

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
vapor density   4.6 (vs air)
vapor pressure   <0.01 mmHg ( 25 °C)
InChI Key   OOCCDEMITAIZTP-QPJJXVBHSA-N
grade   purum
assay   ≥97.0% (GC)
bp   250 °C(lit.)
mp   30-33 °C(lit.)
  30-33 °C
density   1.044 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
GE2200000
Flash Point(F) 
258.8 °F
Flash Point(C) 
126 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?