EMAIL THIS PAGE TO A FRIEND
96340 Sigma-Aldrich

trans-Cinnamic acid

purum, ≥99.0% (T)

Synonym: trans-3-Phenylacrylic acid

  • CAS Number 140-10-3

  • Empirical Formula (Hill Notation) C9H8O2

  • Molecular Weight 148.16

  •  Beilstein Registry Number 1905952

  •  EC Number 205-398-1

  •  MDL number MFCD00004369

  •  PubChem Substance ID 329770497

Purchase

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
GD7850000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?