• USA Home
  • 96458 - Zinc acetate dihydrate

EMAIL THIS PAGE TO A FRIEND
96458 Sigma-Aldrich

Zinc acetate dihydrate

purum p.a., ≥98.0% (KT)

  • CAS Number 5970-45-6

  • Linear Formula Zn(CH3COO)2 · 2H2O

  • Molecular Weight 219.51

  •  Beilstein Registry Number 3732513

  •  EC Number 209-170-2

  •  MDL number MFCD00066961

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Research Essentials, Salts,
InChI Key   BEAZKUGSCHFXIQ-UHFFFAOYSA-L
grade   purum p.a.
assay   ≥98.0% (KT)
pH   6.0-8.0 (25 °C, 50 mg/mL in H2O)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤200 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
ZG8750000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?