• USA Home
  • 96480 - Zinc dimethyldithiocarbamate

EMAIL THIS PAGE TO A FRIEND
96480 Sigma-Aldrich

Zinc dimethyldithiocarbamate

purum, ≥97.0% (KT)

Synonym: Dimethyldithiocarbamic acid zinc salt, Ziram

  • CAS Number 137-30-4

  • Linear Formula (CH3)2NCSSZnSCSN(CH3)2

  • Molecular Weight 305.82

  •  Beilstein Registry Number 3707008

  •  EC Number 205-288-3

  •  MDL number MFCD00064797

  •  PubChem Substance ID 329770513

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Zinc More...
InChI Key   DUBNHZYBDBBJHD-UHFFFAOYSA-L
grade   purum
assay   ≥97.0% (KT)
mp   248-257 °C(lit.)
  248-257 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 2 - EHS - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
ZH0525000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

329711 Zinc dimethyldithiocarbamate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?