• USA Home
  • 96497 - Zinc sulfate heptahydrate

EMAIL THIS PAGE TO A FRIEND
96497 Sigma-Aldrich

Zinc sulfate heptahydrate

tested according to Ph.Eur.

Synonym: Zinci sulfas

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z More...
InChI Key   RZLVQBNCHSJZPX-UHFFFAOYSA-L
assay   ≥99.0%
pharmacopeia   testing conforms to Pharmacopeia
solubility   H2O: 965 g/L at 20 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
ZH5300000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?