• USA Home
  • 96595 - Zirconium(IV) propoxide solution

EMAIL THIS PAGE TO A FRIEND
96595 Sigma-Aldrich

Zirconium(IV) propoxide solution

purum, ~70% in 1-propanol

Synonym: Tetrapropyl zirconate

  • CAS Number 23519-77-9

  • Linear Formula Zr(OCH2CH2CH3)4

  • Molecular Weight 327.57

  •  Beilstein Registry Number 3679533

  •  MDL number MFCD00015307

  •  PubChem Substance ID 329770523

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Zirconium More...
InChI Key   XPGAWFIWCWKDDL-UHFFFAOYSA-N
grade   purum
concentration   ~70% in 1-propanol
refractive index   n20/D 1.454
density   1.058 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
82.4 °F
Flash Point(C) 
28 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

333972 Zirconium(IV) propoxide solution, 70 wt. % in 1-propanol

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?