EMAIL THIS PAGE TO A FRIEND
96710 Sigma-Aldrich

Z-Leu-OH

purum, ≥96.0% (T)

Synonym: N-(Benzyloxycarbonyl)-L-leucine, N-(Benzyloxycarbonyl)leucine, N-(Carbobenzoxy)leucine, N-(Phenylmethoxy)carbonyl-L-leucine, N-Carbobenzoxy-L-leucine, N-Carbobenzyloxy-L-leucine, N-Carboxy-L-leucine N-benzyl ester, N-Cbz-L-leucine, Benzyloxycarbonyl-L-leucine, Benzyloxycarbonylleucine, Carbobenzoxyleucine, L-N-(Benzyloxycarbonyl)leucine, L-(Carbobenzyloxy)leucine, NSC 60039, Z-L-Leucine

  • CAS Number 2018-66-8

  • Linear Formula (CH3)2CHCH2CH(NHCOOCH2C6H5)COOH

  • Molecular Weight 265.30

  •  Beilstein Registry Number 1253861

  •  EC Number 217-960-3

  •  MDL number MFCD00026494

  •  PubChem Substance ID 329770528

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Leucine, Peptide Chemistry More...
InChI Key   USPFMEKVPDBMCG-LBPRGKRZSA-N
grade   purum
assay   ≥96.0% (T)
optical activity   [α]20/D −16±1°, c = 2% in ethanol
refractive index   n20/D 1.512(lit.)
density   1 g/mL at 25 °C(lit.)

Description

Application

Reactant for:
• Peptide-based preparation of potential antitumor agents
• Preparation of cyclopropyl peptidomimetics
• Peptide synthesis

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

521221 Z-Leu-OH, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?