• USA Home
  • 96861 - Piericidin A from Streptomyces mobaraensis

EMAIL THIS PAGE TO A FRIEND
96861 Sigma-Aldrich

Piericidin A from Streptomyces mobaraensis

≥90.0% (HPLC), liquid, green-yellow

Synonym: Shaoguanmycin B

  • CAS Number 2738-64-9

  • Empirical Formula (Hill Notation) C25H37NO4

  • Molecular Weight 415.57

  •  MDL number MFCD01677956

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics N-S, Antibiotics by Application, Antifungal,
InChI Key   BBLGCDSLCDDALX-LKGBESRRSA-N
assay   ≥90.0% (HPLC)
form   liquid
loss   <10% loss on drying
color   green-yellow
shipped in   wet ice
storage temp.   2-8°C

Description

Other Notes

Neurotoxin for the respiratory chain

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3382 6.1 / PGI
WGK Germany 
3
RTECS 
YD4588000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

P4368 Piericidin A from microbial source, >95% (HPLC), DMSO solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?