EMAIL THIS PAGE TO A FRIEND
96915 Sigma-Aldrich

Z-Met-OMe

≥99.0% (sum of enantiomers, HPLC)

Synonym: Z-L-methionine methyl ester

  • CAS Number 56762-93-7

  • Linear Formula C6H5CH2OOCNHCH(COOCH3)CH2CH2SCH3

  • Molecular Weight 297.37

  •  Beilstein Registry Number 2144306

  •  MDL number MFCD00145551

  •  PubChem Substance ID 329770537

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Methionine, Peptide Chemistry More...
InChI Key   YGDBSBBHEWHVAJ-LBPRGKRZSA-N
assay   ≥99.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D −34±2°, c = 1% in methanol
mp   41-44 °C

Description

Other Notes

Intermediate for the synthesis of derivatives of L-vinylglycine [(S)-2-amino-3-butenoic acid]

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?