EMAIL THIS PAGE TO A FRIEND
97025 Sigma-Aldrich

Z-D-phenylalaninol

puriss., ≥99.0% (sum of enantiomers, HPLC)

Synonym: (R)-2-(Z-amino)-3-phenyl-1-propanol

  • CAS Number 58917-85-4

  • Empirical Formula (Hill Notation) C17H19NO3

  • Molecular Weight 285.34

  •  Beilstein Registry Number 2221968

  •  MDL number MFCD00191193

  •  PubChem Substance ID 24890379

Purchase

Properties

Related Categories Amino Acids & Derivatives, Amino Alcohols, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   WPOFMMJJCPZPAO-MRXNPFEDSA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D +43±1°, c = 2% in methanol
mp   92-95 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?