• USA Home
  • 97339 - Cypress oil, from Callitris intratropica

EMAIL THIS PAGE TO A FRIEND
97339 Sigma-Aldrich

Cypress oil, from Callitris intratropica

Australian, natural

Synonym: Cypress oil

Purchase

Properties

quality   natural
refractive index   n20/D 1.507

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
GZ9090000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?