Synonym: Charcoal activated Norit®
CAS Number 7440-44-0
Empirical Formula (Hill Notation) C
Molecular Weight 12.01
Beilstein Registry Number 4360473
EC Number 231-153-3
MDL number MFCD00133992
PubChem Substance ID 24890192
| Related Categories | Activated Charcoal, Activated and Mesoporous Carbon, Adsorbents, Filter Aids and Drying Agents, Alternative Energy, Essential Chemicals, |
| biological source | from wood |
| form | powder |
| quality | chemically activated |
| mfr. no. | Norit CA1 |
| ign. residue | ≤4.0% |
| loss | ≤16% loss on drying |
carrier for catalysts, good filtering properties
Norit is a registered trademark of Norit N.V.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?