• USA Home
  • 98183 - Chloride-selective liquid ion-exchanger microelectrode cocktail A

EMAIL THIS PAGE TO A FRIEND
98183 Sigma-Aldrich

Chloride-selective liquid ion-exchanger microelectrode cocktail A

Selectophore

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophore Cocktails More...
grade   Selectophore

Description

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

91661 Tridodecylmethylammonium chloride, Selectophore
99408 Chloride ionophore I - cocktail A, Selectophore
24894 Chloride ionophore III, Selectophore
24897 Chloride ionophore I, Selectophore
24901 Chloride ionophore II, Selectophore, function tested
92332 Chloride ionophore IV, Selectophore

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?