EMAIL THIS PAGE TO A FRIEND
99053 Sigma-Aldrich

Water

deionized

Purchase

Properties

Related Categories Solvent by Type, Solvents, Water and Water Solutions More...
InChI Key   XLYOFNOQVPJJNP-UHFFFAOYSA-N
quality   deionized
refractive index   n20/D 1.34(lit.)
conductivity   ≤4.3 μS/cm at 20 °C
bp   100 °C(lit.)
density   1.000 g/mL at 3.98 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
ZC0110000
Protocols & Articles

Related Content

Properties of Solvents Table

a. Gant, R., LC/GC 10(7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10mmHg pressure 4. At l=589.32nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?