• USA Home
  • 99110 - Outlet valve No 70501
EMAIL THIS PAGE TO A FRIEND
99110 Sigma-Aldrich
Outlet valve No 70501
steel (bronzed), valve diam. 3 mm
Purchase
Properties
Related Categories Gas Equipment Product Guide, Gas Pressure Tins, Labware More...
material   steel (bronzed)
description   Connecting nut W 21,8 x 1/14 in.
feature   Thread of screw G 1/2 in. with tube connector diam. 8 mm
valve diam.   3 mm
Description
General description
Control valves for cylinders in stainless steel, for corrosive gases, angled, with nut at inlet and unscrewable hose outlet.
Inlet: W 21.8 mm x 14; Hose outlet: G 1/4″ nut.
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?