CAS Number 7757-83-7
Linear Formula Na2SO3
Molecular Weight 126.04
EC Number 231-821-4
MDL number MFCD00003503
PubChem Substance ID 24899438
Popular Documents: Product Information Sheet (PDF)
| Related Categories | Biochemicals and Reagents, Chemical Synthesis, Denaturation, Disulfide Bridge Cleavage Reagents, Inorganic Salts, |
| assay | ≥98% |
2.5 kg in poly bottle
250 g in poly bottle
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?