• USA Home
  • A1056 - Albumin from mouse serum

EMAIL THIS PAGE TO A FRIEND
A1056 Sigma-Aldrich

Albumin from mouse serum

essentially fatty acid free

Synonym: Mouse albumin

Purchase

Properties

Fatty acid content   ~0.005%
storage temp.   2-8°C

Description

Preparation Note

Prepared from Fraction V albumin (A 3139).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A3139 Albumin from mouse serum, lyophilized powder, ≥96% (agarose gel electrophoresis)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?