• USA Home
  • A124 - (−)-N-Allylnormetazocine hydrochloride

EMAIL THIS PAGE TO A FRIEND
A124 Sigma-Aldrich

(−)-N-Allylnormetazocine hydrochloride

analytical standard

Synonym: (−)-NANM, (−)-SKF-10047, (2α,6α)-1,2,3,4,5,6-Hexahydro-6,11-dimethyl-3-(2-propenyl)-2,6-methano-3-benzazocin-8-ol

  • CAS Number 14198-28-8

  • Empirical Formula (Hill Notation) C17H23NO · HCl

  • Molecular Weight 293.83

  •  MDL number MFCD00078553

  •  PubChem Substance ID 24890515

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Structure, Chromatography,
grade   analytical standard
InChI Key   ZTGMHFIGNYXMJV-YGCKJKTASA-N
form   neat
  solid
optical activity   [α]22/D −89.3°, c = 1 in ethanol(lit.)
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
color   white
solubility   H2O: soluble
format   neat

Description

Other Notes

Less active enantiomer.

Same product — same quality — now Sigma-Aldrich brand

Packaging

10 mg in poly bottle

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?