EMAIL THIS PAGE TO A FRIEND
A1772 Sigma-Aldrich

Aluminum oxide

Type WA-4: Acid

Synonym: Alumina

Purchase

Properties

Related Categories Adsorbents, Filter Aids and Drying Agents, Aluminum, Aluminum Oxide, Essential Chemicals, Materials Science,
InChI Key   TWNQGVIAIRXVLR-UHFFFAOYSA-N
type   Type WA-4: Acid
mp   2040 °C(lit.)

Description

Other Notes

Activity Grade Super I

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
BD1200000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?