• USA Home
  • A2018 - Albumin from rat serum

EMAIL THIS PAGE TO A FRIEND
A2018 Sigma-Aldrich

Albumin from rat serum

essentially fatty acid free

Synonym: RSA, Rat albumin

Purchase

Properties

Fatty acid content   ~0.005%
storage temp.   2-8°C

Description

Preparation Note

Prepared from Fraction V albumin (A 6272).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A6414 Albumin from rat serum, lyophilized powder, essentially fatty acid free, essentially globulin free, ≥99% (agarose gel electrophoresis)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?