EMAIL THIS PAGE TO A FRIEND
A2593 Sigma-Aldrich

Azathioprine

meets USP testing specifications

Synonym: 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

Purchase

Properties

Related Categories Analytical/Chromatography, Core Bioreagents, Enzyme Inhibitors, Pharmacopoeia, Pharmacopoeia A-Z,
InChI Key   LMEKQMALGUDUQG-UHFFFAOYSA-N
suitability   meets USP testing specifications
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UO8925000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?