• USA Home
  • A2794 - Acetaminophen solution

EMAIL THIS PAGE TO A FRIEND
A2794 Sigma-Aldrich

Acetaminophen solution

drug standard, 1.0 mg/mL in methanol

Purchase

Properties

grade   drug standard
InChI Key   RZVAJINKPMORJF-UHFFFAOYSA-N
assay   ≥98%
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

A3035 Acetaminophen, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?