EMAIL THIS PAGE TO A FRIEND
A3038 Sigma-Aldrich
Agarose
Type XI, low gelling temperature, suitable for separation of small nucleic acid fragments
Purchase
Properties
type   Type XI
suitability   suitable for separation of small nucleic acid fragments
Description
Analysis Note
The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.
Price and Availability
Suggested Laboratory Gloves
Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Certificate of Origin
Protocols & Articles
Related Content
Agarose Selection Guide
Agarose Selection Guide – Click on the Product Number to Purchase Specialty Agarose – Click on the Product Number to Purchase
Keywords: Cell culture, Electrophoresis, Immunodiffusion, Immunoelectrophoresis, Molecular biology, Protein electrophoresis, Separation
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?