• USA Home
  • A3060 - Albumin from guinea pig serum

EMAIL THIS PAGE TO A FRIEND
A3060 Sigma-Aldrich

Albumin from guinea pig serum

lyophilized powder, essentially fatty acid free

Purchase

Properties

Related Categories Albumin, Albumins from other species, Biochemicals and Reagents, Proteins and Derivatives
form   lyophilized powder
impurities   ~0.005% Fatty acid
storage temp.   2-8°C

Description

Preparation Note

Prepared from Fraction V albumin (A 2639).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?