• USA Home
  • A3183 - Araldite® 506 epoxy resin

EMAIL THIS PAGE TO A FRIEND
A3183 Sigma-Aldrich

Araldite® 506 epoxy resin

Purchase

Properties

Related Categories A, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
composition   Epoxide equivalent weight, 172-185 Da
color   Gardner: ≤3
viscosity   500-700 cP(25 °C)(lit.)

Description

Application

Araldite® 506 epoxy resin has been used as an embedding medium for microscopy.

For embedding samples for microscopy

Packaging

250 g in glass bottle

Quality

A low viscosity modified epoxy resin.

Legal Information

Araldite is a registered trademark of Huntsman Advanced Materials Inc.

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN3082 - class 9 - PG 3 - DOT NA1993 - Environmentally hazardous substances, liquid, n.o.s. HI: all (not BR)
WGK Germany 
2
RTECS 
KC2100000
Flash Point(F) 
485.6 °F
Flash Point(C) 
252 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?