• USA Home
  • A3424 - Bovine Serum Albumin solution

EMAIL THIS PAGE TO A FRIEND
A3424 Sigma-Aldrich

Bovine Serum Albumin solution

30%±2% in 0.85% sodium chloride, aseptically filled, Ultra-high avidity

Synonym: Albumin solution from bovine serum, BSA solution

Purchase

Properties

sterility   aseptically filled
form   solution
contains   0.1% sodium azide as preservative
does not contain   stabilizer
origin   USA origin
concentration   30%±2% in 0.85% sodium chloride
storage temp.   2-8°C

Description

Analysis Note

Tested for avidity with known incomplete antibody (not saline agglutinable).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?