EMAIL THIS PAGE TO A FRIEND
A5409 Sigma-Aldrich

Albumin from hamster

Purchase

Properties

storage temp.   2-8°C

Description

Preparation Note

Fraction V Powder

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?