Synonym: Alumina
CAS Number 1344-28-1
Linear Formula Al2O3
Molecular Weight 101.96
EC Number 215-691-6
MDL number MFCD00003424
PubChem Substance ID 24891058
| Related Categories | Adsorbents for TLC, Adsorbents, Filter Aids and Drying Agents, Alumina & Aluminum Hydroxide TLC Adsorbents, Aluminum, Aluminum Oxide, |
| type | Type CG-20 |
| mp | 2040 °C(lit.) |
| suitability | suitable for chromatography |
1 kg in poly bottle
5 kg in poly drum
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?