Synonym: Glacial acetic acid
CAS Number 64-19-7
Linear Formula CH3CO2H
Molecular Weight 60.05
Beilstein Registry Number 506007
EC Number 200-580-7
MDL number MFCD00036152
PubChem Substance ID 24891075
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | β-Glucuronidase, β-Glucuronidase Drug Analysis Bundle, Acetic Acid, Acetic acid, Acids & Bases, |
| Quality Level | PREMIUM |
| grade | ReagentPlus® |
| vapor density | 2.07 (vs air) |
| assay | ≥99% |
| autoignition temp. | 800 °F |
| expl. lim. | 16 %, 92 °F |
| 4 %, 59 °F |
1, 2.5 L in glass bottle
100, 500 mL in glass bottle
18 L in poly drum
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?