• USA Home
  • A6289 - Albumin from sheep serum

EMAIL THIS PAGE TO A FRIEND
A6289 Sigma-Aldrich

Albumin from sheep serum

essentially fatty acid free

Purchase

Properties

Fatty acid content   ~0.005%
storage temp.   2-8°C

Description

Preparation Note

Prepared from Fraction V albumin (A3264).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A3264 Albumin from sheep serum, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?