EMAIL THIS PAGE TO A FRIEND
A6387 Sigma-Aldrich

Ammonium sulfate

ReagentPlus®, ≥99.0%

Synonym: Mascagnite

Purchase

Properties

Related Categories Ammonium, Ammonium Salts, Chemical Synthesis, Essential Chemicals, Inorganic Salts,
grade   ReagentPlus®
InChI Key   BFNBIHQBYMNNAN-UHFFFAOYSA-N
assay   ≥99.0%
impurities   ≤2 ppm as heavy Metals (Pb)
pH   5.0-6 (25 °C, 132 g/L)
mp   >280 °C (dec.)(lit.)
solubility   H2O: soluble200 gm + 300 mL g/mL
density   1.77 g/mL at 25 °C(lit.)

Description

Preparation Note

Treated to reduce heavy metals

Application

Ammonium sulfate may be used in methyl cellulose precipitable tannin assay to examine wine tannin in finished wine samples. It may also be used in making externally mixed aerosols for analyzing the 1 × 3 tandem differential mobility analyzer (1 × 3-TDMA), which is mainly employed for determining the irreversibility of the hygroscopic growth factor of aerosol particles.

General description

Ammonium sulfate is an ammonium salt commonly used as fertilizer. It can be synthesized by neutralizing sulfuric acid with ammonia. It is reported to enhance the phytotoxicity of several herbicides like glyphosates. It is formed as a co-product during the manufacture of caprolactam, a precursor of nylon.

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Other Notes

This product has been replaced by A5132-Sigma-Aldrich | Ammonium sulfate, ReagentPlus, ≥99.0% | (NH4)2SO4

Price and Availability


Laboratory Gloves

Redi-Dri
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
BS4500000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?