EMAIL THIS PAGE TO A FRIEND
A6597 Sigma-Aldrich

Adaphotris

analytical standard, ≥95% (NMR)

Purchase

Properties

grade   analytical standard
InChI Key   NKBZBQPQAFPKJW-UHFFFAOYSA-M
assay   ≥95% (NMR)
form   neat
  solid
application(s)   qNMR: suitable
format   neat

Description

Biochem/physiol Actions

50 μM inhibits Ca2+/2H+ exchange system of liver mitochondria.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?