Synonym: Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate
CAS Number 12054-85-2
Linear Formula (NH4)6Mo7O24 · 4H2O
Molecular Weight 1235.86
EC Number 234-320-9
MDL number MFCD00167059
PubChem Substance ID 24891201
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | 81.0-83.0% MoO3 basis |
| impurities | ≤0.005% Insoluble matter |
| density | 2.498 g/mL at 25 °C(lit.) |
| anion traces | arsenate, phosphate, and silicate (as SiO2): ≤0.001% |
| chloride (Cl-): ≤0.002% | |
| nitrate (NO3-): passes test (limit about 0.003%) |
1 kg in poly bottle
100, 500 g in poly bottle
2.5 kg in poly drum
May contain darkened ammonium molybdate crystals that are soluble in water.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?