• USA Home
  • A9438 - Albumin from rabbit serum

EMAIL THIS PAGE TO A FRIEND
A9438 Sigma-Aldrich

Albumin from rabbit serum

essentially fatty acid-free

Purchase

Properties

Related Categories Albumin, Albumins from other species, Biochemicals and Reagents, Proteins and Derivatives
quality   essentially fatty acid-free
Fatty acid content   ~0.005%
storage temp.   2-8°C

Description

Preparation Note

Prepared from Fraction V.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?