CAS Number 6484-52-2
Linear Formula NH4NO3
Molecular Weight 80.04
EC Number 229-347-8
MDL number MFCD00011425
PubChem Substance ID 24891481
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Ammonium, Chemical Synthesis, Inorganic Salts, Synthetic Reagents |
| assay | ≥99.0% |
| bp | 210 °C(lit.) |
| mp | 169 °C(lit.) |
2.5 kg in poly drum
500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?