• USA Home
  • B0381 - Bromocresol Green Sultone Form

EMAIL THIS PAGE TO A FRIEND
B0381 Sigma-Aldrich

Bromocresol Green Sultone Form

powder

Synonym: 3′,3′′,5′,5′′-Tetrabromo-m-cresolsulfonephthalein, Bromocresol Blue

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, B, Cell Biology, Hematology and Histology,
InChI Key   FRPHFZCDPYBUAU-UHFFFAOYSA-N
form   powder
pH range   3.8 - 5.4, yellow to blue

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?