• USA Home
  • B168 - (±)-Butaclamol hydrochloride

EMAIL THIS PAGE TO A FRIEND
B168 Sigma-Aldrich

(±)-Butaclamol hydrochloride

analytical standard, for drug analysis

Synonym: AY 23028

  • CAS Number 36504-94-6

  • Empirical Formula (Hill Notation) C25H31NO · HCl

  • Molecular Weight 397.98

  •  MDL number MFCD00082307

  •  PubChem Substance ID 329773194

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antipsychotics Standards, Chemical Structure,
grade   analytical standard, for drug analysis
InChI Key   QZRUMKUMFJJARD-AAJWHBHYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DE8039700

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?