• USA Home
  • B1889 - 4-Benzylamino-7-nitrobenzofurazan

EMAIL THIS PAGE TO A FRIEND
B1889 Sigma-Aldrich

4-Benzylamino-7-nitrobenzofurazan

≥99%

Synonym: 4-Benzylamino-7-nitro-2,1,3-benzoxadiazole, BBD, Benzylamino-NBD

  • CAS Number 18378-20-6

  • Empirical Formula (Hill Notation) C13H10N4O3

  • Molecular Weight 270.24

  •  Beilstein Registry Number 1148439

  •  EC Number 242-261-5

  •  MDL number MFCD00038006

  •  PubChem Substance ID 329773198

Purchase

Properties

InChI Key   GZFKJMWBKTUNJS-UHFFFAOYSA-N
assay   ≥99%
storage temp.   2-8°C

Description

Application

Fluorescent probe of proteins.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?