• USA Home
  • B2906 - Blue rayon trisulfonated

EMAIL THIS PAGE TO A FRIEND
B2906 Sigma-Aldrich

Blue rayon trisulfonated

Synonym: Copper phthalocyanine rayon

Purchase

Description

Other Notes

Rayon linked to trisulfo-copper-phthalocyanine

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?