EMAIL THIS PAGE TO A FRIEND
B3634 Sigma-Aldrich

Benzophenone

Synonym: Diphenyl ketone, Diphenylmethanone, NSC 8077

  • CAS Number 119-61-9

  • Linear Formula (C6H5)2CO

  • Molecular Weight 182.22

  •  Beilstein Registry Number 1238185

  •  EC Number 204-337-6

  •  MDL number MFCD00003076

Purchase

Properties

Related Categories Building Blocks, C13 to C14, Carbonyl Compounds, Chemical Synthesis, Ketones,
vapor pressure   1 mmHg ( 108 °C)
InChI Key   RWCCWEUUXYIKHB-UHFFFAOYSA-N
form   crystalline
bp   305 °C(lit.)
mp   47-51 °C(lit.)
storage temp.   −20°C

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
1
RTECS 
DI9950000
Flash Point(F) 
280.4 °F
Flash Point(C) 
138 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

B9300 Benzophenone, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?